Gene Transcripts

Gene Symbol
Andpro
Entrez Gene ID
25030
Gene Name
androgen regulated protein
Gene Aliases
Crp1, RATANDPRO
Chromosome Location
Chr. 3: 143893005 - 143899609 on Build Rnor_6.0
UniGene ID
Rn.9977
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012718.2 NP_036850.1 1 201
XM_006235184.2 XP_006235246.1 1 440
GenBank mRNA BC065311.1 - 1 181
M27901.1 - 1 185
M58167.1 - 1 202

View Full Product Details