Gene Transcripts
- Gene Symbol
- Irs1
- Entrez Gene ID
- 25467
- Gene Name
- insulin receptor substrate 1
- Gene Aliases
- IRS1IRM
- Chromosome Location
- Chr. 9: 88033668 - 88086488 on Build Rnor_6.0
- UniGene ID
- Rn.10476
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012969.1 |
NP_037101.1
|
1 |
3273 |
|
GenBank mRNA |
X58375.1 |
- |
1 |
3273 |
|
View Full Product Details