Gene Transcripts
- Gene Symbol
- Pax6
- Entrez Gene ID
- 25509
- Gene Name
- paired box 6
- Gene Aliases
- Chromosome Location
- Chr. 3: 95699478 - 95728556 on Build Rnor_6.0
- UniGene ID
- Rn.89724
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013001.2 |
NP_037133.1
|
8, 9 |
926 |
|
XM_006234633.2 |
XP_006234695.1
|
10, 11 |
1351 |
|
XM_006234634.2 |
XP_006234696.1
|
10, 11 |
2807 |
|
XM_006234635.2 |
XP_006234697.1
|
10, 11 |
1266 |
|
XM_006234636.2 |
XP_006234698.1
|
10, 11 |
2257 |
|
XM_008762060.1 |
XP_008760282.1
|
9, 10 |
1450 |
|
GenBank mRNA |
AJ627631.1 |
- |
6, 7 |
808 |
|
AY540905.1 |
- |
7, 8 |
823 |
|
AY540906.1 |
- |
7, 8 |
823 |
|
BC128741.1 |
- |
8, 9 |
926 |
8363963 |
HQ121510.1 |
- |
6, 7 |
809 |
|
U69644.1 |
- |
8, 9 |
1146 |
|
View Full Product Details