Gene Transcripts

Gene Symbol
Rpn1
Entrez Gene ID
25596
Gene Name
ribophorin I
Gene Aliases
RIBI, RPN-I
Chromosome Location
Chr. 4: 119997268 - 120018679 on Build Rnor_6.0
UniGene ID
Rn.233900
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB100593.1 - 3, 4 638
AB100594.1 - 3, 4 638
BC061756.1 - 3, 4 642 5600029
X05300.1 - 3, 4 643

View Full Product Details