Gene Transcripts
- Gene Symbol
- PHOX2B
- Entrez Gene ID
- 8929
- Gene Name
- paired like homeobox 2b
- Gene Aliases
- NBLST2, NBPhox, PMX2B
- Chromosome Location
- Chr. 4: 41744082 - 41748970 on Build GRCh38
- UniGene ID
- Hs.87202
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003924.3 |
NP_003915.2
|
3 |
1910 |
|
GenBank mRNA |
D82344.1 |
- |
3 |
1910 |
|
View Full Product Details