Gene Transcripts
- Gene Symbol
- Mc5r
- Entrez Gene ID
- 25726
- Gene Name
- melanocortin 5 receptor
- Gene Aliases
- Chromosome Location
- Chr. 18: 64115138 - 64116171 on Build Rnor_6.0
- UniGene ID
- Rn.9673
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013182.2 |
NP_037314.1
|
1 |
171 |
|
GenBank mRNA |
L27081.1 |
- |
1 |
383 |
|
View Full Product Details