Gene Transcripts

Gene Symbol
Nhsl2
Entrez Gene ID
100042480
Gene Name
NHS-like 2
Gene Aliases
1110062M06Rik, 6430511F03, AI225852, AW488865, EG621083, ENSMUSG00000073037, Gm10456
Chromosome Location
Chr. X: 101848669 - 102092055 on Build GRCm38
UniGene ID
Mm.232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163610.1 NP_001157082.1 8 6432
GenBank mRNA AK032250.1 - 1 987

View Full Product Details