Gene Transcripts

Gene Symbol
Unc13c
Entrez Gene ID
208898
Gene Name
unc-13 homolog C (C. elegans)
Gene Aliases
1500037O19Rik, AU019458, D9Ertd414e, Munc13-3, Unc13h3
Chromosome Location
Chr. 9: 73479422 - 74008153 on Build GRCm38
UniGene ID
Mm.41035
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081153.1 NP_001074622.1 32 6868
GenBank mRNA AK134584.1 - 6 1065
AK163525.1 - 1 167

View Full Product Details