Gene Transcripts

Gene Symbol
Qrich2
Entrez Gene ID
217341
Gene Name
glutamine rich 2
Gene Aliases
Gm66
Chromosome Location
Chr. 11: 116441323 - 116466269 on Build GRCm38
UniGene ID
Mm.387352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033267.2 NP_001028439.1 9 1289
GenBank mRNA AK131945.1 - 9 1292
BC150976.1 - 9 1142 9087889

View Full Product Details