Gene Transcripts
- Gene Symbol
- Nav3
- Entrez Gene ID
- 260315
- Gene Name
- neuron navigator 3
- Gene Aliases
- 4732483H20Rik, 9630020C08Rik, POMFIL1, Pomfil1p, mKIAA0938, unc53H3
- Chromosome Location
- Chr. 10: 109681573 - 110000391 on Build GRCm38
- UniGene ID
- Mm.225050
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK035945.1 |
- |
|
409 |
|
View Full Product Details