Gene Transcripts

Gene Symbol
Micall1
Entrez Gene ID
27008
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing -like 1
Gene Aliases
260, AI448246, D15Mit260, D15N2e, EST573322, EST820961, MIRab13, mKIAA1668
Chromosome Location
Chr. 15: 79108477 - 79136900 on Build GRCm38
UniGene ID
Mm.196480
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177461.1 NP_803412.1 16 4015
GenBank mRNA AK076226.1 - 3 1316
AK129419.1 - 16 3754
AK147587.1 - 16 3816
AK147646.1 - 16 4028

View Full Product Details