Gene Transcripts

Gene Symbol
Nlrp12
Entrez Gene ID
378425
Gene Name
NLR family, pyrin domain containing 12
Gene Aliases
Nalp12, PYPAF7
Chromosome Location
Chr. 7: 3218643 - 3250159 on Build GRCm38
UniGene ID
Mm.277152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033431.1 NP_001028603.1 3, 4 2254
GenBank mRNA AY364010.1 - 2, 3 1990
BC116253.1 - 3, 4 2127 40093618

View Full Product Details