Gene Transcripts

Gene Symbol
Pabpc1l
Entrez Gene ID
381404
Gene Name
poly(A) binding protein, cytoplasmic 1-like
Gene Aliases
1810053B01Rik, Epab
Chromosome Location
Chr. 2: 164024770 - 164050538 on Build GRCm38
UniGene ID
Mm.82301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114079.2 NP_001107551.1 3, 4 757
GenBank mRNA BC158030.1 - 3, 4 757
EU669908.1 - 3, 4 500

View Full Product Details