Gene Transcripts

Gene Symbol
Susd5
Entrez Gene ID
382111
Gene Name
sushi domain containing 5
Gene Aliases
Gm1126
Chromosome Location
Chr. 9: 114057354 - 114098733 on Build GRCm38
UniGene ID
Mm.376140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101510.1 NP_001094980.1 5 3402
GenBank mRNA AK132299.1 - 5 3324

View Full Product Details