Gene Transcripts

Gene Symbol
Trim80
Entrez Gene ID
432613
Gene Name
tripartite motif-containing 80
Gene Aliases
4933422H20Rik, Trim47l
Chromosome Location
Chr. 11: 115440545 - 115448268 on Build GRCm38
UniGene ID
Mm.308822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033775.3 NP_001028947.1 2 95
GenBank mRNA AK133415.1 - 2 96
BC147608.1 - 2 113 9056598
BC157912.1 - 2 113 9088129

View Full Product Details