Gene Transcripts

Gene Symbol
Kprp
Entrez Gene ID
433619
Gene Name
keratinocyte expressed, proline-rich
Gene Aliases
1110001M24Rik, AA589586
Chromosome Location
Chr. 3: 92823074 - 92827247 on Build GRCm38
UniGene ID
Mm.28359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028629.1 NP_082905.1 2 2134
GenBank mRNA AK003253.1 - 1 424
BC141372.1 - 2 2117 9055985

View Full Product Details