Gene Transcripts

Gene Symbol
Skint1
Entrez Gene ID
639781
Gene Name
selection and upkeep of intraepithelial T cells 1
Gene Aliases
OTTMUSG00000008542, Skint-1
Chromosome Location
Chr. 4: 112006269 - 112029538 on Build GRCm38
UniGene ID
Mm.398873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102662.1 NP_001096132.1 5 886
GenBank mRNA EF494889.1 - 5 886
EU099296.1 - 5 886

View Full Product Details