Gene Transcripts

Gene Symbol
Krtdap
Entrez Gene ID
64661
Gene Name
keratinocyte differentiation associated protein
Gene Aliases
Kdap, sk221
Chromosome Location
Chr. 7: 30787905 - 30791083 on Build GRCm38
UniGene ID
Mm.268157
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033131.3 NP_001028303.1 6 340
GenBank mRNA AK131603.1 - 6 341
BC132510.1 - 6 340 40130787
BC132512.1 - 6 340 40130789

View Full Product Details