Gene Transcripts

Gene Symbol
Krtap4-1
Entrez Gene ID
665891
Gene Name
keratin associated protein 4-1
Gene Aliases
A030010K20Rik
Chromosome Location
Chr. 11: 99627229 - 99628239 on Build GRCm38
UniGene ID
Mm.394141
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048196.1 NP_001041661.1 1 656
GenBank mRNA AK137752.1 - 1 657
BC147087.1 - 1 627 9056137
BC147088.1 - 1 627 9056139

View Full Product Details