Gene Transcripts

Gene Symbol
Krtap4-9
Entrez Gene ID
665998
Gene Name
keratin associated protein 4-9
Gene Aliases
OTTMUSG00000002198
Chromosome Location
Chr. 11: 99785200 - 99786257 on Build GRCm38
UniGene ID
Mm.425699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085548.2 NP_001079017.1 1 21

View Full Product Details