Gene Transcripts

Gene Symbol
Myeov2
Entrez Gene ID
66915
Gene Name
myeloma overexpressed 2
Gene Aliases
1110002M09Rik
Chromosome Location
Chr. 1: 92637145 - 92641985 on Build GRCm38
UniGene ID
Mm.30179
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163425.1 NP_001156897.1 3 207
GenBank mRNA AA764377.1 - 24
AK003315.1 - 3 188
AK012198.1 - 3 180
AK152318.1 - 3 174
BC049759.1 - 3 207
BC147334.1 - 3 193 9056392
BC147335.1 - 3 193 9056395
BC147659.1 - 3 193 9087448
BC147669.1 - 3 193 9087458
BY631936.1 - 3 202

View Full Product Details