Gene Transcripts

Gene Symbol
Sdhaf1
Entrez Gene ID
68332
Gene Name
succinate dehydrogenase complex assembly factor 1
Gene Aliases
0610010E21Rik, AI430885, AW490662, Lyrm8
Chromosome Location
Chr. 7: 30321409 - 30322375 on Build GRCm38
UniGene ID
Mm.6844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033140.3 NP_001028312.2 1 464
GenBank mRNA AK002462.1 - 1 446
AK155437.1 - 1 428
CA451543.1 - 1 436

View Full Product Details