Gene Transcripts

Gene Symbol
Chrnb2
Entrez Gene ID
54239
Gene Name
cholinergic receptor, nicotinic, beta 2 (neuronal)
Gene Aliases
Chromosome Location
Chr. 2: 189088570 - 189096785 on Build Rnor_6.0
UniGene ID
Rn.53978
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019297.1 NP_062170.1 4 484
GenBank mRNA AY574258.1 - 4 296
L31622.1 - 4 484

View Full Product Details