Gene Transcripts
- Gene Symbol
- Mcpt4
- Entrez Gene ID
- 54270
- Gene Name
- mast cell protease 4
- Gene Aliases
- Chromosome Location
- Chr. 15: 34751550 - 34754409 on Build Rnor_6.0
- UniGene ID
- Rn.10698
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019321.2 |
NP_062194.1
|
4 |
546 |
|
GenBank mRNA |
U67907.1 |
- |
4 |
548 |
|
View Full Product Details