Gene Transcripts
- Gene Symbol
- Cym
- Entrez Gene ID
- 56825
- Gene Name
- chymosin
- Gene Aliases
- Chromosome Location
- Chr. 2: 209962848 - 209973205 on Build Rnor_6.0
- UniGene ID
- Rn.48676
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020091.1 |
NP_064476.1
|
6, 7 |
782 |
|
GenBank mRNA |
AJ251688.1 |
- |
6, 7 |
782 |
|
View Full Product Details