Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002016.2 | NP_001002016.2 | 1, 2 | 562 | |
GenBank mRNA | DN934590.1 | - | 1, 2 | 302 | |
FM031779.1 | - | 1, 2 | 554 | ||
X66870.1 | - | 1, 2 | 521 | ||
X76297.1 | - | 1, 2 | 284 |