Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022198.2 | NP_071534.2 | 8, 9 | 1101 | |
XM_006256828.2 | XP_006256890.1 | 8, 9 | 1404 | ||
GenBank mRNA | BC092209.1 | - | 8, 9 | 1101 | |
FQ210965.1 | - | 8, 9 | 1140 | ||
Z36944.1 | - | 6, 7 | 809 |