Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022515.1 | NP_071960.1 | 5 | 367 | |
GenBank mRNA | BC058473.1 | - | 5 | 392 | |
FQ209984.1 | - | 5 | 386 | ||
FQ220432.1 | - | 5 | 396 | ||
FQ221142.1 | - | 5 | 385 | ||
FQ221221.1 | - | 5 | 392 | ||
FQ221486.1 | - | 5 | 388 | ||
FQ221683.1 | - | 5 | 379 | ||
FQ221688.1 | - | 5 | 388 | ||
FQ221810.1 | - | 5 | 388 | ||
FQ221946.1 | - | 5 | 388 | ||
FQ221988.1 | - | 5 | 392 | ||
FQ222823.1 | - | 5 | 388 | ||
FQ223282.1 | - | 5 | 388 | ||
FQ224199.1 | - | 5 | 388 | ||
FQ224575.1 | - | 5 | 392 | ||
FQ224676.1 | - | 5 | 388 | ||
X78443.1 | - | 5 | 367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006257178.2 | XP_006257240.1 | 2 | 380 |