Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031102.2 | NP_112364.1 | 3 | 172 | |
XM_006229161.2 | XP_006229223.1 | 3 | 653 | ||
GenBank mRNA | BC084727.1 | - | 2 | 164 | |
CB577548.1 | - | 3 | 172 | ||
DQ403026.1 | - | 2 | 88 | ||
FQ211765.1 | - | 3 | 190 | ||
FQ221110.1 | - | 3 | 190 | ||
FQ222651.1 | - | 3 | 190 | ||
FQ228615.1 | - | 3 | 193 | ||
M20156.1 | - | 2 | 158 |