Gene Transcripts
- Gene Symbol
- Il3
- Entrez Gene ID
- 24495
- Gene Name
- interleukin 3
- Gene Aliases
- Chromosome Location
- Chr. 10: 39620549 - 39622973 on Build Rnor_6.0
- UniGene ID
- Rn.10652
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031513.1 |
NP_113701.1
|
2 |
205 |
|
GenBank mRNA |
U81492.1 |
- |
2 |
205 |
|
View Full Product Details