Gene Transcripts

Gene Symbol
Pax4
Entrez Gene ID
83630
Gene Name
paired box 4
Gene Aliases
Chromosome Location
Chr. 4: 55735643 - 55753635 on Build Rnor_6.0
UniGene ID
Rn.14531
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031799.1 NP_113987.1 4 463
XM_006236231.2 XP_006236293.1 5 6516
XM_006236232.2 XP_006236294.1 6 1512
XM_006236233.2 XP_006236295.1 6 954
GenBank mRNA AF053100.1 - 4 463
AF053101.1 - 4 463
AF053102.1 - 4 463
AF053103.1 - 4 463
AF198155.1 - 3 415
AF198156.1 - 3 415

View Full Product Details