Gene Transcripts
- Gene Symbol
- Kcnma1
- Entrez Gene ID
- 83731
- Gene Name
- potassium channel, calcium activated large conductance subfamily M alpha, member 1
- Gene Aliases
- BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
- Chromosome Location
- Chr. 15: 344204 - 1048849 on Build Rnor_6.0
- UniGene ID
- Rn.30616
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031828.1 |
NP_114016.1
|
1, 2 |
884 |
|
GenBank mRNA |
AF083341.1 |
- |
1, 2 |
320 |
|
AF135265.1 |
- |
1, 2 |
738 |
|
AY330290.1 |
- |
1, 2 |
209 |
|
AY330291.1 |
- |
1, 2 |
209 |
|
AY330293.2 |
- |
1, 2 |
209 |
|
AY330294.1 |
- |
1, 2 |
209 |
|
AY344964.1 |
- |
1, 2 |
209 |
|
AY344965.2 |
- |
1, 2 |
713 |
|
U40603.1 |
- |
1, 2 |
884 |
|
U55995.1 |
- |
1, 2 |
362 |
|
U93052.1 |
- |
1, 2 |
310 |
|
View Full Product Details