Gene Transcripts

Gene Symbol
Nploc4
Entrez Gene ID
140639
Gene Name
nuclear protein localization 4 homolog (S. cerevisiae)
Gene Aliases
Npl4
Chromosome Location
Chr. 10: 109553518 - 109610087 on Build Rnor_6.0
UniGene ID
Rn.217306
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080577.2 NP_542144.1 14 1535
GenBank mRNA AF234600.1 - 14 1463
BC101887.1 - 14 1535 7930738

View Full Product Details