Gene Transcripts

Gene Symbol
Slc9a5
Entrez Gene ID
192215
Gene Name
solute carrier family 9, subfamily A (NHE5, cation proton antiporter 5), member 5
Gene Aliases
Nhe5
Chromosome Location
Chr. 19: 37305248 - 37325335 on Build Rnor_6.0
UniGene ID
Rn.24105
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138858.1 NP_620213.1 4 756
GenBank mRNA AF100172.1 - 4 756

View Full Product Details