Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138855.1 | NP_620210.1 | 6 | 795 | |
XM_006239766.1 | XP_006239828.1 | 6 | 1029 | ||
XM_006239767.2 | XP_006239829.1 | 6 | 993 | ||
XM_006239768.2 | XP_006239830.1 | 6 | 860 | ||
XM_008764502.1 | XP_008762724.1 | 6 | 1695 | ||
GenBank mRNA | AF492385.1 | - | 6 | 795 | |
BC078729.1 | - | 5 | 718 | 7134092 |