Gene Transcripts
- Gene Symbol
- Slc9a5
- Entrez Gene ID
- 192215
- Gene Name
- solute carrier family 9, subfamily A (NHE5, cation proton antiporter 5), member 5
- Gene Aliases
- Nhe5
- Chromosome Location
- Chr. 19: 37305248 - 37325335 on Build Rnor_6.0
- UniGene ID
- Rn.24105
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138858.1 |
NP_620213.1
|
15 |
2166 |
|
GenBank mRNA |
AF100172.1 |
- |
15 |
2166 |
|
View Full Product Details