Gene Transcripts

Gene Symbol
Nupl1
Entrez Gene ID
245922
Gene Name
nucleoporin like 1
Gene Aliases
p58/p45
Chromosome Location
Chr. 15: 40496752 - 40545929 on Build Rnor_6.0
UniGene ID
Rn.11099
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139091.2 NP_620791.1 2 283
XM_006252093.2 XP_006252155.1 2 388
GenBank mRNA AF000900.1 - 2 220
AF000901.1 - 1 176
BC085690.1 - 2 283 7190650
U63839.1 - 2 296

View Full Product Details