Gene Transcripts

Gene Symbol
Gata4
Entrez Gene ID
54254
Gene Name
GATA binding protein 4
Gene Aliases
Chromosome Location
Chr. 15: 46386703 - 46432914 on Build Rnor_6.0
UniGene ID
Rn.26251
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144730.1 NP_653331.1 3 1206
XM_006252189.2 XP_006252251.1 2 786
GenBank mRNA L22761.1 - 3 1206

View Full Product Details