Gene Transcripts
- Gene Symbol
- Gja8
- Entrez Gene ID
- 29601
- Gene Name
- gap junction protein, alpha 8
- Gene Aliases
- Cx50
- Chromosome Location
- Chr. 2: 199050854 - 199052470 on Build Rnor_6.0
- UniGene ID
- Rn.86658
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153465.2 |
NP_703195.2
|
1 |
552 |
|
View Full Product Details