Gene Transcripts
- Gene Symbol
- Zp4
- Entrez Gene ID
- 282833
- Gene Name
- zona pellucida glycoprotein 4
- Gene Aliases
- Zp-4
- Chromosome Location
- Chr. 17: 65497359 - 65503912 on Build Rnor_6.0
- UniGene ID
- Rn.91141
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172330.3 |
NP_758833.2
|
7, 8 |
1032 |
|
GenBank mRNA |
AF456325.1 |
- |
7, 8 |
1033 |
|
View Full Product Details