Gene Transcripts
- Gene Symbol
- PLXDC1
- Entrez Gene ID
- 57125
- Gene Name
- plexin domain containing 1
- Gene Aliases
- TEM3, TEM7
- Chromosome Location
- Chr. 17: 39063303 - 39151649 on Build GRCh38
- UniGene ID
- Hs.125036
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020405.4 |
NP_065138.2
|
2, 3 |
478 |
|
GenBank mRNA |
AF279144.2 |
- |
2, 3 |
348 |
|
AF378753.1 |
- |
2, 3 |
348 |
|
AK093589.1 |
- |
2, 3 |
499 |
|
AK127539.1 |
- |
1, 2 |
295 |
|
AK303699.1 |
- |
1, 2 |
165 |
|
AK312999.1 |
- |
2, 3 |
300 |
|
AY704670.1 |
- |
2, 3 |
348 |
|
AY704671.1 |
- |
2, 3 |
348 |
|
AY704672.1 |
- |
2, 3 |
294 |
|
BC036059.1 |
- |
2, 3 |
444 |
5298499 |
DA205327.1 |
- |
2, 3 |
478 |
|
KU257674.1 |
- |
2, 3 |
206 |
|
View Full Product Details