Gene Transcripts

Gene Symbol
Lim2
Entrez Gene ID
114903
Gene Name
lens intrinsic membrane protein 2
Gene Aliases
MP20, Mp19
Chromosome Location
Chr. 1: 98495082 - 98501248 on Build Rnor_6.0
UniGene ID
Rn.9688
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053771.1 NP_446223.1 2, 3 215
GenBank mRNA L04191.1 - 2, 3 193
M87053.1 - 2, 3 215
S55224.1 - 2, 3 206

View Full Product Details