Gene Transcripts

Gene Symbol
Gosr2
Entrez Gene ID
64154
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
Chromosome Location
Chr. 10: 91736779 - 91756081 on Build Rnor_6.0
UniGene ID
Rn.13518
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031685.2 NP_113873.1 3 177
GenBank mRNA AF007549.1 - 3 191
BC061994.1 - 3 177 5599557
U91539.1 - 3 176

View Full Product Details