Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301353.1 | NP_001288282.1 | 6 | 957 | |
NM_001301354.1 | NP_001288283.1 | 5 | 891 | ||
NM_007470.4 | NP_031496.2 | 6 | 961 | ||
GenBank mRNA | AK135046.1 | - | 6 | 749 | |
AK157917.1 | - | 6 | 749 | ||
AK157925.1 | - | 6 | 748 | ||
AK158118.1 | - | 6 | 745 | ||
AK158405.1 | - | 6 | 751 | ||
AK160729.1 | - | 5 | 680 | ||
AK162392.1 | - | 6 | 749 | ||
BC145907.1 | - | 4 | 638 | 40131210 | |
BC145909.1 | - | 4 | 638 | 40131212 | |
X82648.1 | - | 5 | 695 |