Gene Transcripts

Gene Symbol
Nkx3-2
Entrez Gene ID
12020
Gene Name
NK3 homeobox 2
Gene Aliases
Bapx1, NKX3.2, Nkx-3.2
Chromosome Location
Chr. 5: 41761483 - 41764220 on Build GRCm38
UniGene ID
Mm.2530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007524.3 NP_031550.2 2 1301
GenBank mRNA BC145872.1 - 2 1296
BC145874.1 - 2 1296 40131177
U87957.1 - 2 1301

View Full Product Details