Gene Transcripts
- Gene Symbol
- Cd48
- Entrez Gene ID
- 12506
- Gene Name
- CD48 antigen
- Gene Aliases
- AI449234, AW610730, BCM1, BLAST, BLAST-1, BLAST1, Bcm-1, MEM-102, SLAMF2, Sgp-60
- Chromosome Location
- Chr. 1: 171681421 - 171705257 on Build GRCm38
- UniGene ID
- Mm.1738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007649.4 |
NP_031675.1
|
1, 2 |
120 |
|
GenBank mRNA |
AB196820.1 |
- |
1, 2 |
81 |
|
AB196821.1 |
- |
1, 2 |
81 |
|
AB196822.1 |
- |
1, 2 |
81 |
|
AB196823.1 |
- |
1, 2 |
81 |
|
AK007741.1 |
- |
1, 2 |
121 |
|
AK084984.1 |
- |
1, 2 |
149 |
|
AK088027.1 |
- |
1, 2 |
167 |
|
AK146173.1 |
- |
1, 2 |
167 |
|
AK151601.1 |
- |
1, 2 |
111 |
|
BC060977.1 |
- |
1, 2 |
148 |
30296173 |
X17501.1 |
- |
1, 2 |
131 |
|
X53526.1 |
- |
1, 2 |
131 |
|
View Full Product Details