Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008185.3 | NP_032211.3 | 3, 4 | 447 | |
GenBank mRNA | AK002338.1 | - | 3, 4 | 447 | |
BC012254.1 | - | 3, 4 | 362 | 4193816 | |
BC055020.1 | - | 3, 4 | 383 | 4161144 | |
W54467.1 | - | 3, 4 | 398 | ||
X98055.1 | - | 3, 4 | 359 |