Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008605.3 | NP_032631.3 | 3 | 455 | |
GenBank mRNA | AK089295.1 | - | 424 | ||
AK089309.1 | - | 3 | 424 | ||
AK089335.1 | - | 3 | 420 | ||
AK089452.1 | - | 3 | 424 | ||
AK089509.1 | - | 4 | 298 | ||
AK089523.1 | - | 3 | 424 | ||
AK090051.1 | - | 3 | 426 | ||
AK150596.1 | - | 3 | 424 | ||
AK151021.1 | - | 3 | 424 | ||
AK151273.1 | - | 3 | 424 | ||
AK151933.1 | - | 3 | 424 | ||
AK155139.1 | - | 3 | 423 | ||
AK155743.1 | - | 3 | 424 | ||
AK155747.1 | - | 3 | 424 | ||
AK155748.1 | - | 3 | 424 | ||
AK155841.1 | - | 3 | 482 | ||
AK155865.1 | - | 3 | 423 | ||
AK170360.1 | - | 3 | 424 | ||
AK170364.1 | - | 3 | 424 | ||
AK170500.1 | - | 4 | 298 | ||
AK170667.1 | - | 2 | 1471 | ||
AK170798.1 | - | 3 | 423 | ||
AK170804.1 | - | 3 | 423 | ||
AK170976.1 | - | 3 | 424 | ||
AK171006.1 | - | 3 | 424 | ||
AK171032.1 | - | 3 | 423 | ||
AK171036.1 | - | 3 | 424 | ||
AK171176.1 | - | 3 | 424 | ||
AK171428.1 | - | 3 | 424 | ||
AK171435.1 | - | 2 | 1471 | ||
AK171447.1 | - | 3 | 424 | ||
AK171448.1 | - | 3 | 424 | ||
AK171449.1 | - | 3 | 424 | ||
AK171450.1 | - | 3 | 424 | ||
AK171466.1 | - | 3 | 424 | ||
AK171468.1 | - | 3 | 424 | ||
AK171470.1 | - | 3 | 424 | ||
AK171472.1 | - | 3 | 424 | ||
AK171485.1 | - | 3 | 424 | ||
AK171518.1 | - | 3 | 424 | ||
AK171542.1 | - | 3 | 424 | ||
AK171570.1 | - | 3 | 424 | ||
AY622972.1 | - | 1 | 21 | ||
BC019135.1 | - | 3 | 418 | 5037674 | |
CT010377.1 | - | 3 | 371 | ||
M82831.1 | - | 3 | 371 |