Gene Transcripts

Gene Symbol
Odf1
Entrez Gene ID
18285
Gene Name
outer dense fiber of sperm tails 1
Gene Aliases
Chromosome Location
Chr. 15: 38219203 - 38226735 on Build GRCm38
UniGene ID
Mm.252830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008757.3 NP_032783.2 2 654
GenBank mRNA BC145758.1 - 2 654
EU791469.1 - 2 654
X79446.1 - 2 658

View Full Product Details