Gene Transcripts

Gene Symbol
Pdcd1
Entrez Gene ID
18566
Gene Name
programmed cell death 1
Gene Aliases
Ly101, PD-1, Pdc1
Chromosome Location
Chr. 1: 94038305 - 94052553 on Build GRCm38
UniGene ID
Mm.5024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008798.2 NP_032824.1 2 379
GenBank mRNA AK039828.1 - 2 359
BC119179.1 - 2 348 8733928
BC120602.1 - 2 348 40129525
X67914.1 - 2 379

View Full Product Details